SY3H3D6803AE Synthonix Corporation

1-Methyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylic acid

  • Cas Number : 32286-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50188.html

Safety Information

MSDS https://synthonix.com/50188.html
Availability details for
Quantity
  • Availability
  • Add to Cart