SY3H3D6803AF Synthonix Corporation

Methyl 5,6,7,8-tetrahydroimidazo[1,5-a]pyridine-6-carboxylate

  • MDL number MFCD22417057
  • Cas Number : 139183-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50190.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/50190.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart