SY3H3D6803CE Synthonix Corporation

Methyl 5-(tert-butyl)isoxazole-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 517870-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50378.html

Safety Information

MSDS https://synthonix.com/50378.html
Availability details for
Quantity
  • Availability
  • Add to Cart