SY3H3D6803CF Synthonix Corporation

2-(2-Methoxyethoxy)benzenesulfonamide

  • MDL number No MDL found
  • Cas Number : 82031-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50381.html

Safety Information

MSDS https://synthonix.com/50381.html
Availability details for
Quantity
  • Availability
  • Add to Cart