SY3H3D6803D0 Synthonix Corporation

3-Morpholino-5,6-dihydropyridin-2(1H)-one

  • MDL number No MDL found
  • Cas Number : 545445-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50403.html

Safety Information

MSDS https://synthonix.com/50403.html
Availability details for
Quantity
  • Availability
  • Add to Cart