SY3H3D6803D8 Synthonix Corporation

2-(4-Methylpiperazin-1-yl)pyrimidin-4-amine

  • MDL number No MDL found
  • Cas Number : 57005-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50462.html

Safety Information

MSDS https://synthonix.com/50462.html
Availability details for
Quantity
  • Availability
  • Add to Cart