SY3H3D6803D9 Synthonix Corporation

2-(4-Methylpiperazin-1-yl)pyrimidin-5-amine

  • MDL number No MDL found
  • Cas Number : 943757-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50463.html

Safety Information

MSDS https://synthonix.com/50463.html
Availability details for
Quantity
  • Availability
  • Add to Cart