SY3H3D6803DA Synthonix Corporation

6-(4-Methylpiperazin-1-yl)pyridazin-3-amine

  • MDL number No MDL found
  • Cas Number : 66346-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50464.html

Safety Information

MSDS https://synthonix.com/50464.html
Availability details for
Quantity
  • Availability
  • Add to Cart