SY3H3D6803E2 Synthonix Corporation

Methyl 2-((tert-butoxycarbonyl)(methyl)amino)acetate

  • MDL number No MDL found
  • Cas Number : 42492-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50549.html

Safety Information

MSDS https://synthonix.com/50549.html
Availability details for
Quantity
  • Availability
  • Add to Cart