SY3H3D6803E6 Synthonix Corporation

(S)-Methyl 2-amino-3-((tert-butoxycarbonyl)amino)propanoate

  • Cas Number : 77087-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/50577.html

Safety Information

MSDS https://synthonix.com/50577.html
Availability details for
Quantity
  • Availability
  • Add to Cart