SY3H3D6803E8 Synthonix Corporation

Methyl 3-(trimethylsilyl)pent-4-enoate

  • MDL number No MDL found
  • Cas Number : 185411-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50583.html

Safety Information

MSDS https://synthonix.com/50583.html
Availability details for
Quantity
  • Availability
  • Add to Cart