SY3H3D6803E9 Synthonix Corporation

4-(2-Methoxyethoxy)cyclohexanol

  • Cas Number : 107025-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50585.html

Safety Information

MSDS https://synthonix.com/50585.html
Availability details for
Quantity
  • Availability
  • Add to Cart