SY3H3D6803EB Synthonix Corporation

Methyl 3-((3-(dimethylamino)propyl)amino)propanoate

  • MDL number No MDL found
  • Cas Number : 90796-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50612.html

Safety Information

MSDS https://synthonix.com/50612.html
Availability details for
Quantity
  • Availability
  • Add to Cart