SY3H3D6803F0 Synthonix Corporation

Methyl 2,6-dichloro-4-cyanobenzoate

  • MDL number No MDL found
  • Cas Number : 409127-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/50752.html

Safety Information

MSDS https://synthonix.com/50752.html
Availability details for
Quantity
  • Availability
  • Add to Cart