SY3H3D6803F3 Synthonix Corporation

Methyl 3-cyano-5-nitrobenzoate

  • MDL number No MDL found
  • Cas Number : 99066-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/50832.html

Safety Information

MSDS https://synthonix.com/50832.html
Availability details for
Quantity
  • Availability
  • Add to Cart