SY3H3D6803F6 Synthonix Corporation

4-Methyl-3-(trifluoromethyl)benzonitrile

  • MDL number No MDL found
  • Cas Number : 261952-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/50859.html

Safety Information

MSDS https://synthonix.com/50859.html
Availability details for
Quantity
  • Availability
  • Add to Cart