SY3H3D6803FB Synthonix Corporation

Methyl 6-bromobenzo[d]thiazole-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 1187928-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/50871.html

Safety Information

MSDS https://synthonix.com/50871.html
Availability details for
Quantity
  • Availability
  • Add to Cart