SY3H3D6803FD Synthonix Corporation

Methyl 4-chloro-2-cyanobenzoate

  • MDL number No MDL found
  • Cas Number : 58331-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50873.html

Safety Information

MSDS https://synthonix.com/50873.html
Availability details for
Quantity
  • Availability
  • Add to Cart