SY3H3D6803FF Synthonix Corporation

Methyl 2-cyano-4-fluorobenzoate

  • Cas Number : 127510-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50881.html

Safety Information

MSDS https://synthonix.com/50881.html
Availability details for
Quantity
  • Availability
  • Add to Cart