SY3H3D680400 Synthonix Corporation

Methyl 2-cyano-5-fluorobenzoate

  • MDL number No MDL found
  • Cas Number : 606080-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50882.html

Safety Information

MSDS https://synthonix.com/50882.html
Availability details for
Quantity
  • Availability
  • Add to Cart