SY3H3D680402 Synthonix Corporation

Methyl 4-cyano-2-fluorobenzoate

  • MDL number No MDL found
  • Cas Number : 175596-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/50884.html

Safety Information

MSDS https://synthonix.com/50884.html
Availability details for
Quantity
  • Availability
  • Add to Cart