SY3H3D680403 Synthonix Corporation

Methyl 4-cyano-3-fluorobenzoate

  • Cas Number : 268734-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50885.html

Safety Information

MSDS https://synthonix.com/50885.html
Availability details for
Quantity
  • Availability
  • Add to Cart