SY3H3D680406 Synthonix Corporation

Methyl 5-chlorobenzo[d]oxazole-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 27383-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50898.html

Safety Information

MSDS https://synthonix.com/50898.html
Availability details for
Quantity
  • Availability
  • Add to Cart