SY3H3D680410 Synthonix Corporation

Methyl 3,5-difluoro-4-formylbenzoate

  • MDL number No MDL found
  • Cas Number : 1415124-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/50998.html

Safety Information

MSDS https://synthonix.com/50998.html
Availability details for
Quantity
  • Availability
  • Add to Cart