SY3H3D680417 Synthonix Corporation

Methyl 3-bromoimidazo[1,2-a]pyridine-6-carboxylate

  • MDL number No MDL found
  • Cas Number : 886361-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51103.html

Safety Information

MSDS https://synthonix.com/51103.html
Availability details for
Quantity
  • Availability
  • Add to Cart