SY3H3D68041A Synthonix Corporation

Methyl 4-fluoropyrazolo[1,5-a]pyridine-3-carboxylate

  • Cas Number : 1802489-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51114.html

Safety Information

MSDS https://synthonix.com/51114.html
Availability details for
Quantity
  • Availability
  • Add to Cart