SY3H3D68041B Synthonix Corporation

Methyl 6-fluoropyrazolo[1,5-a]pyridine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 1802489-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/51115.html

Safety Information

MSDS https://synthonix.com/51115.html
Availability details for
Quantity
  • Availability
  • Add to Cart