SY3H3D68041C Synthonix Corporation

6-Methoxy-2-(trifluoromethyl)-1H-benzo[d]imidazole

  • Cas Number : 3671-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51132.html

Safety Information

MSDS https://synthonix.com/51132.html
Availability details for
Quantity
  • Availability
  • Add to Cart