SY3H3D680427 Synthonix Corporation

2-Methylbenzo[d]oxazole-6-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 864274-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51187.html

Safety Information

MSDS https://synthonix.com/51187.html
Availability details for
Quantity
  • Availability
  • Add to Cart