SY3H3D68042E Synthonix Corporation

5-Methoxyisoindoline-1,3-dione

  • MDL number No MDL found
  • Cas Number : 50727-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51205.html

Safety Information

MSDS https://synthonix.com/51205.html
Availability details for
Quantity
  • Availability
  • Add to Cart