SY3H3D680430 Synthonix Corporation

Methyl benzo[d]oxazole-2-carboxylate

  • Cas Number : 27383-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51214.html

Safety Information

MSDS https://synthonix.com/51214.html
Availability details for
Quantity
  • Availability
  • Add to Cart