SY3H3D680433 Synthonix Corporation

Methyl benzo[d]oxazole-7-carboxylate

  • MDL number No MDL found
  • Cas Number : 1086378-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51217.html

Safety Information

MSDS https://synthonix.com/51217.html
Availability details for
Quantity
  • Availability
  • Add to Cart