SY3H3D680434 Synthonix Corporation

2-Methylbenzo[d]oxazole-5-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 90322-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51218.html

Safety Information

MSDS https://synthonix.com/51218.html
Availability details for
Quantity
  • Availability
  • Add to Cart