SY3H3D680436 Synthonix Corporation

6-Methoxy-1H-benzo[d][1,3]oxazine-2,4-dione

  • MDL number No MDL found
  • Cas Number : 37795-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51225.html

Safety Information

MSDS https://synthonix.com/51225.html
Availability details for
Quantity
  • Availability
  • Add to Cart