SY3H3D68043A Synthonix Corporation

2-(Methoxycarbonyl)-6-nitrobenzoic acid

  • Cas Number : 21606-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51232.html

Safety Information

MSDS https://synthonix.com/51232.html
Availability details for
Quantity
  • Availability
  • Add to Cart