SY3H3D68043D Synthonix Corporation

Methyl 4-(bromomethyl)-2,5-difluorobenzoate

  • MDL number No MDL found
  • Cas Number : 1355488-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51253.html

Safety Information

MSDS https://synthonix.com/51253.html
Availability details for
Quantity
  • Availability
  • Add to Cart