SY3H3D68043F Synthonix Corporation

2-Methyl-5-(trifluoromethyl)benzoic acid

  • Cas Number : 13055-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51265.html

Safety Information

MSDS https://synthonix.com/51265.html
Availability details for
Quantity
  • Availability
  • Add to Cart