SY3H3D680446 Synthonix Corporation

Methyl 2-(2-bromophenyl)-2-oxoacetate

  • MDL number MFCD18206246
  • Cas Number : 122394-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Colorless to light-yellow to yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51277.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51277.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart