SY3H3D680449 Synthonix Corporation

Methyl 3-bromo-4-(difluoromethoxy)benzoate

  • MDL number No MDL found
  • Cas Number : 200956-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51281.html

Safety Information

MSDS https://synthonix.com/51281.html
Availability details for
Quantity
  • Availability
  • Add to Cart