SY3H3D68044A Synthonix Corporation

Methyl 5-bromo-2-(difluoromethoxy)benzoate

  • MDL number No MDL found
  • Cas Number : 1131587-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/51282.html

Safety Information

MSDS https://synthonix.com/51282.html
Availability details for
Quantity
  • Availability
  • Add to Cart