SY3H3D680450 Synthonix Corporation

Methyl 3-(trifluoromethoxy)benzoate

  • MDL number No MDL found
  • Cas Number : 148438-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/51288.html

Safety Information

MSDS https://synthonix.com/51288.html
Availability details for
Quantity
  • Availability
  • Add to Cart