SY3H3D680455 Synthonix Corporation

4-methylsulfonyl-2-(trifluoromethyl)benzaldehyde

  • MDL number No MDL found
  • Cas Number : 1215310-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51298.html

Safety Information

MSDS https://synthonix.com/51298.html
Availability details for
Quantity
  • Availability
  • Add to Cart