SY3H3D680459 Synthonix Corporation

5-Methyl-4-quinazolone

  • Cas Number : 75844-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51344.html

Safety Information

MSDS https://synthonix.com/51344.html
Availability details for
Quantity
  • Availability
  • Add to Cart