SY3H3D68045B Synthonix Corporation

7-Methylquinazolin-4(3H)-one

  • MDL number No MDL found
  • Cas Number : 75844-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/51356.html

Safety Information

MSDS https://synthonix.com/51356.html
Availability details for
Quantity
  • Availability
  • Add to Cart