SY3H3D68045E Synthonix Corporation

6-Methylimidazo[1,2-a]pyridine-2-carbaldehyde

  • MDL number MFCD05863353
  • Cas Number : 202348-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51366.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51366.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart