SY3H3D680463 Synthonix Corporation

6-Methoxyquinoxalin-2(1H)-one

  • MDL number No MDL found
  • Cas Number : 91192-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51375.html

Safety Information

MSDS https://synthonix.com/51375.html
Availability details for
Quantity
  • Availability
  • Add to Cart