SY3H3D680469 Synthonix Corporation

Methyl 2H-indazole-7-carboxylate

  • MDL number No MDL found
  • Cas Number : 952479-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51385.html

Safety Information

MSDS https://synthonix.com/51385.html
Availability details for
Quantity
  • Availability
  • Add to Cart