SY3H3D68046D Synthonix Corporation

6-Methylquinazoline-2,4(1H,3H)-dione

  • MDL number No MDL found
  • Cas Number : 62484-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51390.html

Safety Information

MSDS https://synthonix.com/51390.html
Availability details for
Quantity
  • Availability
  • Add to Cart