SY3H3D68046F Synthonix Corporation

Methyl 2-(3-cyanopyridin-4-yl)acetate

  • MDL number No MDL found
  • Cas Number : 124870-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/51393.html

Safety Information

MSDS https://synthonix.com/51393.html
Availability details for
Quantity
  • Availability
  • Add to Cart