SY3H3D680472 Synthonix Corporation

Methyl imidazo[1,5-a]pyridine-6-carboxylate

  • MDL number No MDL found
  • Cas Number : 139183-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51397.html

Safety Information

MSDS https://synthonix.com/51397.html
Availability details for
Quantity
  • Availability
  • Add to Cart